Learn More
(1S)-3-(Dimethylamino)-1-(thiophen-2-yl)propan-1-ol, Mikromol™
Description
(1S)-3-(Dimethylamino)-1-(thiophen-2-yl)propan-1-ol
Specifications
Specifications
| Chemical Name or Material | (S)-1-beta-Hydroxy-1-(2-thienyl)-3-dimethylaminopropane |
| CAS | 132335-44-5 |
| Analyte or Component Names | (S)-1-beta-Hydroxy-1-(2-thienyl)-3-dimethylaminopropane |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H15 N O S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Thiophenemethanol, α-[2-(dimethylamino)ethyl]-, (αS)-, 2-Thiophenemethanol, α-[2-(dimethylamino)ethyl]-, (S)-, (αS)-α-[2-(Dimethylamino)ethyl]-2-thiophenemethanol, (1S)-3-(Dimethylamino)-1-(2-thienyl)-1-propanol, (S)-(-)-3-(Dimethylamino)-1-(thiophen-2-yl)propan-1-ol, (S)-(-)-N,N-Dimethyl-3-hydroxy-3-(2-thienyl)propanamine, (S)-3-(Dimethylamino)-1-(2-thienyl)-1-propanol, (S)-N,N-Dimethyl-3-(2-thienyl)-3-hydroxypropanamine, (S)-N,N-Dimethyl-3-hydroxy-3-(2-thienyl)-1-propylamine |
| InChI Formula | InChI=1S/C9H15NOS/c1-10(2)6-5-8(11)9-4-3-7-12-9/h3-4,7-8,11H,5-6H2,1-2H3/t8-/m0/s1 |
| SMILES | CN(C)CC[C@H](O)c1cccs1 |
| IUPAC Name | (1S)-3-(dimethylamino)-1-thiophen-2-ylpropan-1-ol |
| Show More |