missing translation for 'onlineSavingsMsg'
Läs mer

(1S,3R)-1-Benzo[1,3]dioxol-5-yl-2-(2-chloro-acetyl)-2,3,4,9-tetrahydro-1H-b-carboline-3-carboxylic Acid Methyl Ester, TRC

Produktkod. 30134706
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30134706 2.5 g 2.5g
17919204 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30134706 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1217002.5G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 629652-40-0
Molekylformel C22H19ClN2O5
Molekylvikt (g/mol) 426.85
Synonym (1S,3R)-1-(1,3-Benzodioxol-5-yl)-2-(2-chloroacetyl)-2,3,4,9-tetrahydro1H-pyrido[3,4-b]indole-3-carboxylic Acid Methyl Ester, (1S,3R)-1-(1,3-Benzodioxol-5-yl)-2-(chloroacetyl)-2,3,4,9-tetrahydro1H-pyrido[3,4-b]indole-3-carboxylic Acid Methyl Ester;
IUPAC-namn methyl (1S,3R)-1-(1,3-benzodioxol-5-yl)-2-(2-chloroacetyl)-1,3,4,9-tetrahydropyrido[3,4-b]indole-3-carboxylate
LEDER O=C(N1[C@@H](C2=CC=C3C(OCO3)=C2)C(NC4=CC=CC=C54)=C5C[C@@H]1C(OC)=O)CCl

Specifikationer

Kemiskt namn eller material (1S,3R)-1-Benzo[1,3]dioxol-5-yl-2-(2-chloro-acetyl)-2,3,4,9-tetrahydro-1H-b-carboline-3-carboxylic Acid Methyl Ester
Analyt- eller komponentnamn (1S,3R)-1-Benzo[1,3]dioxol-5-yl-2-(2-chloro-acetyl)-2,3,4,9-tetrahydro-1H-b-carboline-3-carboxylic Acid Methyl Ester
Frakt skick Room Temperature
InChI formel InChI=1S/C22H19ClN2O5/c1-28-22(27)16-9-14-13-4-2-3-5-15(13)24-20(14)21(25(16)19(26)10-23)12-6-7-17-18(8-12)30-11-29-17/h2-8,16,21,24H,9-11H2,1H3/t16-,21+/m1/s1
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 2.5 g
Formel vikt 426.0982
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.