Läs mer
(1S,3S)-1-(1,3-Benzodioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic Acid Methyl Ester, TRC
Produktkod. 30339712
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30339712 25 mg
1 options
Produktkod. 30339712 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB20002025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1S,3S)-1-(1,3-Benzodioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic Acid Methyl Ester
CAS 171596-43-3
Molekylformel C20H18N2O4
Kvantitet 25 mg
Synonym (1S-cis)-1-(1,3-benzodioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic Acid Methyl Ester
InChI formel InChI=1S/C20H18N2O4/c1-24-20(23)15-9-13-12-4-2-3-5-14(12)21-19(13)18(22-15)11-6-7-16-17(8-11)26-10-25-16/h2-8,15,18,21-22H,9-10H2,1H3/t15-,18-/m0/s1
LEDER COC(=O)[C@@H]1CC2=C([C@@H](N1)C3=CC4=C(C=C3)OCO4)NC5=CC=CC=C25
IUPAC-namn methyl (1S,3S)-1-(1,3-benzodioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate
Molekylvikt (g/mol) 350.37
Formel vikt 350.1267