Läs mer
(1S,3S)-Solifenacin Hydrochloride, TRC
Produktkod. 17970745
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17970745 25 mg -
17972305 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17970745 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS67671525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 180468-38-6
Molekylformel C23 H26 N2 O2 . Cl H
Molekylvikt (g/mol) 398.9257
Synonym 2(1H)-Isoquinolinecarboxylic acid, 3,4-dihydro-1-phenyl-, (3S)-1-azabicyclo[2.2.2]oct-3-yl ester, hydrochloride (1:1), (1S)-, 2(1H)-Isoquinolinecarboxylic acid, 3,4-dihydro-1-phenyl-, (3S)-1-azabicyclo[2.2.2]oct-3-yl ester, monohydrochloride, (1S)- (9CI), 2(1H)-Isoquinolinecarboxylic acid, 3,4-dihydro-1-phenyl-, 1-azabicyclo[2.2.2]oct-3-yl ester, monohydrochloride, [S-(R*,R*)]-, (1S,3'S)-Solifenacin hydrochloride, Solifenacin S,S-diastereomer, (S)-Quinuclidin-3-yl (S)-1-phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate hydrochloride, (1S)-3,4-Dihydro-1-phenyl-2(1H)-isoquinolinecarboxylic Acid (3S)-1-Azabicyclo-[2.2.2]oct-3-yl Ester Hydrochloride
IUPAC-namn [(3S)-1-azabicyclo[2.2.2]octan-3-yl] (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate;hydrochloride
LEDER Cl.O=C(O[C@@H]1CN2CCC1CC2)N3CCc4ccccc4[C@@H]3c5ccccc5
Specifikationer
Kemiskt namn eller material (1S,3S)-Solifenacin Hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C23H26N2O2.ClH/c26-23(27-21-16-24-13-10-18(21)11-14-24)25-15-12-17-6-4-5-9-20(17)22(25)19-7-2-1-3-8-19;/h1-9,18,21-22H,10-16H2;1H/t21-,22+;/m1./s1
Formel vikt 398.1761