Läs mer
(1S,4R)-4-Amino-2-cyclopentene-1-carboxylic Acid Methyl Ester L-Tartrate, TRC
Produktkod. 30323126
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30323126 50 mg
1 options
Produktkod. 30323126 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60387550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1S,4R)-4-Amino-2-cyclopentene-1-carboxylic Acid Methyl Ester L-Tartrate
CAS 419563-22-7
Molekylformel C11H17NO8
Kvantitet 50 mg
Synonym (1S,4R)-4-Amino-2-cyclopentene-1-carboxylic Acid Methyl Ester (2R,3R)-2,3-Dihydroxybutanedioate
InChI formel InChI=1S/C7H11NO2.C4H6O6/c1-10-7(9)5-2-3-6(8)4-5;5-1(3(7)8)2(6)4(9)10/h2-3,5-6H,4,8H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t5-,6+;1-,2-/m11/s1
LEDER COC(=O)[C@H]1C[C@@H](N)C=C1.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC-namn (2R,3R)-2,3-dihydroxybutanedioic acid;methyl (1S,4R)-4-aminocyclopent-2-ene-1-carboxylate
Molekylvikt (g/mol) 291.25
Formel vikt 291.0954