Läs mer
(1S,AlphaS)-trans-Cypermethrin, TRC
Produktkod. 30358732
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30358732 10 mg -
30354000 25 mg -
30366087 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30358732 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98900810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 83860-31-5
Molekylformel C22H19Cl2NO3
Molekylvikt (g/mol) 416.3
Synonym (S)-Cyano(3-phenoxyphenyl)methyl(1S,3R)-3-(2,2-dichloroethenyl)-2,2-dimethyl-cyclopropanecarboxylate
IUPAC-namn [(S)-cyano-(3-phenoxyphenyl)methyl] (1S,3R)-3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate
LEDER CC1(C)[C@@H](C=C(Cl)Cl)[C@@H]1C(=O)O[C@H](C#N)c2cccc(Oc3ccccc3)c2
Specifikationer
Kemiskt namn eller material (1S,alphaS)-trans-Cypermethrin
Kvantitet 10 mg
InChI formel InChI=1S/C22H19Cl2NO3/c1-22(2)17(12-19(23)24)20(22)21(26)28-18(13-25)14-7-6-10-16(11-14)27-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/t17-,18+,20+/m0/s1
Formel vikt 415.0742