Läs mer
(1S-cis)-3-(2-Chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethylcyclopropanecarboxylic Acid (R)-Alpha-Methylbenzenemethanamine, TRC
Produktkod. 30364373
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364373 500 mg
30365461 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30364373 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC422065500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (1S-cis)-3-(2-Chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethylcyclopropanecarboxylic Acid (R)-alpha-Methylbenzenemethanamine
CAS 90191-32-5
Molekylformel C17H21ClF3NO2
Kvantitet 500 mg
Synonym (R)-α-Methylbenzenemethanamine (1S-cis)-3-(2-Chloro-3,3,3-trifluoro-1-propenyl)-2,2-dimethylcyclopropanecarboxylate
InChI formel InChI=1S/C9H10ClF3O2.C8H11N/c1-8(2)4(6(8)7(14)15)3-5(10)9(11,12)13;1-7(9)8-5-3-2-4-6-8/h3-4,6H,1-2H3,(H,14,15);2-7H,9H2,1H3/t4-,6-;7-/m11/s1
LEDER [C@H](C)(N)C1=CC=CC=C1.C(=C(C(F)(F)F)Cl)[C@@H]1[C@H](C(O)=O)C1(C)C
IUPAC-namn (R)-1-phenylethan-1-amine (1S,3S)-3-(2-chloro-3,3,3-trifluoroprop-1-en-1-yl)-2,2-dimethylcyclopropane-1-carboxylate
Molekylvikt (g/mol) 363.8
Formel vikt 363.12