missing translation for 'onlineSavingsMsg'
Learn More

2-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetamide, Mikromol™

Product Code. 30051044
Change view
Click to view available options
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30051044 100 mg
30052166 25 mg
30461199 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30051044 Supplier Mikromol Supplier No. MM0084.20

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetamide
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetamide
CAS 30763-03-2
Analyt- eller komponentnamn 2-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetamide
Frakt skick Room Temperature
Molekylformel C9 H8 N2 O4 S
Certifikat och efterlevnad ISO 17025
Synonym 2-(1,1-Dioxido-3-oxo-1,2-benzisothiazol-2(3H)-yl)acetamide
InChI formel InChI=1S/C9H8N2O4S/c10-8(12)5-11-9(13)6-3-1-2-4-7(6)16(11,14)15/h1-4H,5H2,(H2,10,12)
LEDER NC(=O)CN1C(=O)c2ccccc2S1(=O)=O
IUPAC-namn 2-(1,1,3-trioxo-1,2-benzothiazol-2-yl)acetamide
Molekylvikt (g/mol) 240.24
Kvantitet 100 mg
Formel vikt 240.0205 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.