Läs mer
2-(1,2,3,4-Tetrahydronaphthalen-1-yl)-1H-imidazole, TRC
Produktkod. 30425967
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425967 25 mg
30411408 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425967 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29411025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 945389-41-3
Molekylformel C13H14N2
Molekylvikt (g/mol) 198.26
Synonym 2-(1,2,3,4-Tetrahydro-1-naphthalenyl)-1H-imidazole
IUPAC-namn 2-(1,2,3,4-tetrahydronaphthalen-1-yl)-1H-imidazole
LEDER C1CC(c2ccccc2C1)c3ncc[nH]3
Specifikationer
Kemiskt namn eller material 2-(1,2,3,4-Tetrahydronaphthalen-1-yl)-1H-imidazole
Kvantitet 25 mg
InChI formel InChI=1S/C13H14N2/c1-2-6-11-10(4-1)5-3-7-12(11)13-14-8-9-15-13/h1-2,4,6,8-9,12H,3,5,7H2,(H,14,15)
Formel vikt 198.1157