Läs mer
2-(1,3-Benzothiazol-2-ylthio)succinic Acid, TRC
Produktkod. 30136027
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136027 500 mg
30136881 100 mg
30005446 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136027 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB130343500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 95154-01-1
Molekylformel C11 H9 N O4 S2
Molekylvikt (g/mol) 283.32
Synonym (2-Benzothiazolylthio)succinic Acid, Butanedioic acid, 2-(2-benzothiazolylthio)-, Butanedioic acid, (2-benzothiazolylthio)- (9CI), 2-(2-Benzothiazolylthio)butanedioic acid, (2-Benzothiazolylthio)butanedioic acid, (2-Benzothiazolylthio)succinic acid, 2-(Benzo[d]thiazol-2-ylthio)succinic acid, 2-(Benzo[d]thiazol-2-ylthio)succinicacid, 2-(Benzothiazol-2-ylsulfanyl)succinic acid, Benzthiazol-2-ylthio succinic acid, Halox 650, Irgacor 252, Irgacor 252LD, Reocor 152
IUPAC-namn 2-(1,3-benzothiazol-2-ylsulfanyl)butanedioic acid
LEDER OC(=O)CC(Sc1nc2ccccc2s1)C(=O)O
Specifikationer
Kemiskt namn eller material 2-(1,3-Benzothiazol-2-ylthio)succinic Acid
Kvantitet 500 mg
InChI formel InChI=1S/C11H9NO4S2/c13-9(14)5-8(10(15)16)18-11-12-6-3-1-2-4-7(6)17-11/h1-4,8H,5H2,(H,13,14)(H,15,16)
Formel vikt 282.9973