Läs mer
2-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetyl]-butanedioic Acid Diethyl Ester, TRC
Produktkod. 30373139
Change view
Kvantitet:
2.5 mg
Produktkod. Kvantitet
30373139 2.5 mg
1 options
Produktkod. 30373139 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4491082.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetyl]-butanedioic Acid Diethyl Ester
CAS 125428-01-5
Molekylformel C18H19NO7
Kvantitet 2.5 mg
Synonym 2-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetyl]-butanedioic Acid Diethyl Ester, diethyl 2-(2-(1,3-dioxoisoindolin-2-yl)acetyl)succinate
InChI formel InChI=1S/C18H19NO7/c1-3-25-15(21)9-13(18(24)26-4-2)14(20)10-19-16(22)11-7-5-6-8-12(11)17(19)23/h5-8,13H,3-4,9-10H2,1-2H3
LEDER O=C(N1CC(C(C(OCC)=O)CC(OCC)=O)=O)C(C=CC=C2)=C2C1=O
IUPAC-namn 2-[(1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetyl]-butanedioic Acid Diethyl Ester
Molekylvikt (g/mol) 361.35
Formel vikt 361.12