Läs mer
2-((1-Benzyl-1,2,3,6-tetrahydropyridin-4-yl)methyl)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one (>90%), TRC
Produktkod. 30342381
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30342381 100 mg
30332744 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30342381 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB296135100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-((1-Benzyl-1,2,3,6-tetrahydropyridin-4-yl)methyl)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one
CAS 608511-44-0
Molekylformel C24H27NO3
Kvantitet 100 mg
Synonym 1-Benzyl-4-[(5,6-dimethoxy-1-oxoindan-2-yl)methyl]-1,2,3,6-tetrahydropyridine, 2,3-Dihydro-5,6-dimethoxy-2-[[1,2,3,6-tetrahydro-1-(phenylmethyl)-4-pyridinyl]methyl]-1H-inden-1-one
InChI formel InChI=1S/C24H27NO3/c1-27-22-14-19-13-20(24(26)21(19)15-23(22)28-2)12-17-8-10-25(11-9-17)16-18-6-4-3-5-7-18/h3-8,10,14-15,17,20H,9,11-13,16H2,1-2H3
LEDER COc1cc2CC(CC3CCN(Cc4ccccc4)C=C3)C(=O)c2cc1OC
IUPAC-namn 2-[(1-benzyl-3,4-dihydro-2H-pyridin-4-yl)methyl]-5,6-dimethoxy-2,3-dihydroinden-1-one
Molekylvikt (g/mol) 377.48
Formel vikt 377.1991