Läs mer
2-(1-Hydroxyethyl) Promazine Sulfoxide (mixture of diastereomers), TRC
Produktkod. 30010670
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010670 2.5 mg 2.5mg
30133382 100 mg -
30006504 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010670 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9422902.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73644-42-5
Molekylformel C19H24N2O2S
Molekylvikt (g/mol) 344.47
Synonym 2-(1-Hydroxyethyl) Promazine Sulfoxide
IUPAC-namn 1-[10-[3-(dimethylamino)propyl]-5-oxophenothiazin-2-yl]ethanol
LEDER CC(O)c1ccc2c(c1)N(CCCN(C)C)c3ccccc3S2=O
Specifikationer
Kemiskt namn eller material 2-(1-Hydroxyethyl) Promazine Sulfoxide
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C19H24N2O2S/c1-14(22)15-9-10-19-17(13-15)21(12-6-11-20(2)3)16-7-4-5-8-18(16)24(19)23/h4-5,7-10,13-14,22H,6,11-12H2,1-3H3
Formel vikt 344.1558
Procent renhet >95%