Läs mer
2-(1-Methyl-1H-pyrazol-3-yl)ethanamine, TRC
Produktkod. 30480703
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480703 100 mg
30483654 10 mg
30465021 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30480703 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM324273100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1221824-87-8
Molekylformel C6H11N3
Molekylvikt (g/mol) 125.17
IUPAC-namn 2-(1-methylpyrazol-3-yl)ethanamine
LEDER Cn1ccc(CCN)n1
Specifikationer
Kemiskt namn eller material 2-(1-Methyl-1H-pyrazol-3-yl)ethanamine
Kvantitet 100 mg
InChI formel InChI=1S/C6H11N3/c1-9-5-3-6(8-9)2-4-7/h3,5H,2,4,7H2,1H3
Formel vikt 125.0953