Läs mer
2-(1-Methyl-1H-pyrazol-4-yl)ethanamine (~90%), TRC
Produktkod. 30476588
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476588 10 mg
30482300 100 mg
30480845 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476588 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32806810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 796845-58-4
Molekylformel C6H11N3
Molekylvikt (g/mol) 125.17
Synonym 2-(1-Methyl-1H-pyrazol-4-yl)ethanamine
IUPAC-namn 2-(1-methylpyrazol-4-yl)ethanamine
LEDER Cn1cc(CCN)cn1
Specifikationer
Kemiskt namn eller material 2-(1-Methyl-1H-pyrazol-4-yl)ethanamine
Kvantitet 10 mg
InChI formel InChI=1S/C6H11N3/c1-9-5-6(2-3-7)4-8-9/h4-5H,2-3,7H2,1H3
Formel vikt 125.0953