Läs mer
2-(1-Methylethyl)-2-propylpropanedioic Acid 1,3-Diethyl Ester, TRC
Produktkod. 30469203
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30469203 100 mg
1 options
Produktkod. 30469203 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM305005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(1-Methylethyl)-2-propylpropanedioic Acid 1,3-Diethyl Ester
CAS 62391-98-4
Molekylformel C13H24O4
Kvantitet 100 mg
Synonym Diethyl 2-propan-2-yl-2-propylpropanedioate, Diethyl 2-isopropyl-2-propyl-propanedioate, 2-Isopropyl-2-propylpropanedioic Acid Diethyl Ester, 2-Isopropyl-2-propyl-malonic Acid Diethyl Ester, Diethyl 2-propan-2-yl-2-propyl-propanedioate, Propanedioic Acid','(1-methylethyl)propyl-','diethyl Ester, Diethyl 2-isopropyl-2-propylmalonate
InChI formel InChI=1S/C13H24O4/c1-6-9-13(10(4)5,11(14)16-7-2)12(15)17-8-3/h10H,6-9H2,1-5H3
LEDER CCCC(C(C)C)(C(=O)OCC)C(=O)OCC
IUPAC-namn diethyl 2-propan-2-yl-2-propylpropanedioate
Molekylvikt (g/mol) 244.33
Formel vikt 244.1675