Learn More
Alfa Aesar™ 2-(1-Naphthyl)pyrrolidine, 95%
Brand:  Alfa Aesar™ H61800.03
Additional Details : CAS Number : 121193-91-7 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15448378
  • 4184.41 SEK / 1g
Estimated Shipment date
from Supplier 15-11-2018
Add to basket
Description
Description
Applications
2-(1-Naphthyl)pyrrolidine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Chemical Identifiers
121193-91-7
197.281
FEJHFYHZVJWOOE-UHFFFAOYSA-N
249758
C1CC(NC1)C2=CC=CC3=CC=CC=C32
C14H15N
MFCD02663484
2-naphthalen-1-yl pyrrolidine, 2-naphthalen-1-yl-pyrrolidine, 2-1-naphthyl pyrrolidine, pyrrolidine,2-1-naphthalenyl, acmc-20ct66
2-naphthalen-1-ylpyrrolidine
Specifications
2-(1-Naphthyl)pyrrolidine
1g
C14H15N
2-naphthalen-1-yl pyrrolidine, 2-naphthalen-1-yl-pyrrolidine, 2-1-naphthyl pyrrolidine, pyrrolidine,2-1-naphthalenyl, acmc-20ct66
C1CC(NC1)C2=CC=CC3=CC=CC=C32
197.281
197.28
95%
Yellow
121193-91-7
MFCD02663484
FEJHFYHZVJWOOE-UHFFFAOYSA-N
2-naphthalen-1-ylpyrrolidine
249758