Läs mer
2-(1-Naphthyloxy)propanoic Acid, TRC
Produktkod. 30472604
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30472604 1 g
1 options
Produktkod. 30472604 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN1651011G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(1-Naphthyloxy)propanoic Acid
CAS 13949-67-2
Molekylformel C13 H12 O3
Kvantitet 1 g
Synonym Propionic acid, 2-(1-naphthyloxy)- (6CI,8CI), 2-(1-Naphthalenyloxy)propanoic acid, 2-(1-Naphthyloxy)propanoic acid, NSC 523899, Propanoic acid, 2-(1-naphthalenyloxy)-, 2-(1-Naphthyloxy)-Propionic Acid (NOPA), NOPA, 2-(1-Naphthalenyloxy)propanoic Acid, NSC 523899, Propanoic acid, 2-(1-naphthalenyloxy)-
InChI formel InChI=1S/C13H12O3/c1-9(13(14)15)16-12-8-4-6-10-5-2-3-7-11(10)12/h2-9H,1H3,(H,14,15)
LEDER CC(Oc1cccc2ccccc12)C(=O)O
IUPAC-namn 2-naphthalen-1-yloxypropanoic acid
Molekylvikt (g/mol) 216.23
Formel vikt 216.0786