Learn More
2-(1-Pyrrolidinyl)phenol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Insoluble in water.
Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Insoluble in water.
Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | 109°C to 112°C |
Quantity | 1 g |
Beilstein | 135437 |
Solubility Information | Insoluble in water. |
Formula Weight | 163.22 |
Percent Purity | 98% |
Chemical Name or Material | 2-(1-Pyrrolidinyl)phenol |