Läs mer
2-(1H-Indazol-6-ylsulfinyl)-N-methylbenzamide, TRC
Produktkod. 30407065
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30407065 50 mg
30404973 100 mg
30405532 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30407065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI62810250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1959572-97-4
Molekylformel C15H13N3O2S
Molekylvikt (g/mol) 299.35
Synonym Axitinib Impurity C
IUPAC-namn 2-(1H-indazol-6-ylsulfinyl)-N-methylbenzamide
LEDER CNC(=O)c1ccccc1S(=O)c2ccc3cn[nH]c3c2
Specifikationer
Kemiskt namn eller material 2-(1H-Indazol-6-ylsulfinyl)-N-methylbenzamide
Kvantitet 50 mg
InChI formel InChI=1S/C15H13N3O2S/c1-16-15(19)12-4-2-3-5-14(12)21(20)11-7-6-10-9-17-18-13(10)8-11/h2-9H,1H3,(H,16,19)(H,17,18)
Formel vikt 299.0728