Läs mer
2-(1H-Pyrazol-4-yl)aniline, TRC
Produktkod. 30340539
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30340539 50 mg
30337654 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30340539 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB53020050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(1H-Pyrazol-4-yl)aniline
CAS 1342757-33-8
Molekylformel C9H9N3
Kvantitet 50 mg
Synonym 2-(1H-pyrazol-4-yl)aniline dihydrochloride
InChI formel InChI=1S/C9H9N3/c10-9-4-2-1-3-8(9)7-5-11-12-6-7/h1-6H,10H2,(H,11,12)
LEDER Nc1ccccc1c2cn[nH]c2
IUPAC-namn 2-(1H-pyrazol-4-yl)aniline
Molekylvikt (g/mol) 159.18
Formel vikt 159.0796