Läs mer
2'-(1H-Tetrazol-5-yl)-1,1'-biphenyl-4-carboxaldehyde (Losartan Impurity), TRC
Produktkod. 30455571
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30455571 25 mg -
30452986 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30455571 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT30851025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 151052-40-3
Molekylformel C14 H10 N4 O
Molekylvikt (g/mol) 250.255
Synonym 2'-(2H-Tetrazol-5-yl)-[1,1'-biphenyl]-4-carboxaldehyde
IUPAC-namn 4-[2-(1H-tetrazol-5-yl)phenyl]benzaldehyde
LEDER O=Cc1ccc(cc1)c2ccccc2c3nnn[nH]3
Specifikationer
Kemiskt namn eller material 2'-(1H-Tetrazol-5-yl)-1,1'-biphenyl-4-carboxaldehyde (Losartan Impurity)
Kvantitet 25 mg
InChI formel InChI=1S/C14H10N4O/c19-9-10-5-7-11(8-6-10)12-3-1-2-4-13(12)14-15-17-18-16-14/h1-9H,(H,15,16,17,18)
Formel vikt 250.085