Läs mer
2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Desethyl Etodolac), Mikromol™
Produktkod. 30461236
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461236 4 x 25 mg
30050069 25 mg
30052437 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30461236 Leverantör Mikromol Leverantörsnummer MM0911.0100254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Desethyl Etodolac), Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Desethyl Etodolac)
CAS 41339-67-7
Analyt- eller komponentnamn 2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Desethyl Etodolac)
Frakt skick Room Temperature
Molekylformel C15 H17 N O3
Certifikat och efterlevnad ISO 9001
Synonym Etodolac Imp. A (EP), 2-[(1RS)-1-Ethyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid, 8-Desethyl Etodolac, Etodolac Impurity A
InChI formel InChI=1S/C15H17NO3/c1-2-15(9-13(17)18)14-11(7-8-19-15)10-5-3-4-6-12(10)16-14/h3-6,16H,2,7-9H2,1H3,(H,17,18)
Lösningstyp Neat
LEDER CCC1(CC(=O)O)OCCc2c1[nH]c3ccccc23
IUPAC-namn 2-(1-ethyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetic acid
Molekylvikt (g/mol) 259.3
Kvantitet 4 x 25 mg
Formel vikt 259.1208 g/mol