Läs mer
2-[(1RS)-1-Ethyl-8-methyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Methyl Etodolac), Mikromol™
Produktkod. 18081820
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18081820 25 mg
18023820 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18081820 Leverantör Mikromol Leverantörsnummer MM0911.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[(1RS)-1-Ethyl-8-methyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (8-Methyl Etodolac)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 8-Methyl Etodolac
CAS 41340-19-6
Analyt- eller komponentnamn 8-Methyl Etodolac
Frakt skick Room Temperature
Molekylformel C16 H19 N O3
Certifikat och efterlevnad ISO 17025
Synonym Etodolac Imp. B (EP), 2-[(1RS)-1-Ethyl-8-methyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid, 8-Methyl Etodolac, Etodolac Impurity B
InChI formel InChI=1S/C16H19NO3/c1-3-16(9-13(18)19)15-12(7-8-20-16)11-6-4-5-10(2)14(11)17-15/h4-6,17H,3,7-9H2,1-2H3,(H,18,19)
LEDER CCC1(CC(=O)O)OCCc2c1[nH]c3c(C)cccc23
IUPAC-namn 2-(1-ethyl-8-methyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetic acid
Molekylvikt (g/mol) 273.33
Kvantitet 25 mg
Formel vikt 273.1365 g/mol