Läs mer
2-[[(1RS)-1-Methyl-2-[4-(trifluoromethyl)phenyl]ethyl]amino]ethyl Benzoate Hydrochloride, Mikromol™
Produktkod. 30049722
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30049722 25 mg
1 options
Produktkod. 30049722 Leverantör Mikromol Leverantörsnummer MM1298.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[[(1RS)-1-Methyl-2-[4-(trifluoromethyl)phenyl]ethyl]amino]ethyl Benzoate Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[[(1RS)-1-Methyl-2-[4-(trifluoromethyl)phenyl]ethyl]amino]ethyl Benzoate Hydrochloride
CAS 22444-70-8
Analyt- eller komponentnamn 2-[[(1RS)-1-Methyl-2-[4-(trifluoromethyl)phenyl]ethyl]amino]ethyl Benzoate Hydrochloride
Frakt skick Room Temperature
Molekylformel C19 H20 F3 N O2 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Benfluorex Hydrochloride Imp. B (EP), Benfluorex Imp. B (EP), 2-[[(1RS)-1-Methyl-2-[4-(trifluoromethyl)phenyl]ethyl]amino]ethyl Benzoate Hydrochloride, Benfluorex Hydrochloride Imp. B (EP) as Hydrochloride, Benfluorex Hydrochloride Impurity B as Hydrochloride, Benfluorex Impurity B as Hydrochloride
InChI formel InChI=1S/C19H20F3NO2.ClH/c1-14(13-15-7-9-17(10-8-15)19(20,21)22)23-11-12-25-18(24)16-5-3-2-4-6-16;/h2-10,14,23H,11-13H2,1H3;1H
LEDER Cl.CC(Cc1ccc(cc1)C(F)(F)F)NCCOC(=O)c2ccccc2
IUPAC-namn 2-[1-[4-(trifluoromethyl)phenyl]propan-2-ylamino]ethyl benzoate;hydrochloride
Molekylvikt (g/mol) 387.82
Kvantitet 25 mg
Formel vikt 387.1213 g/mol