Läs mer
2-[(1RS)-8-Ethyl-1-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (1-Propyl Etodolac), Mikromol™
Produktkod. 30049718
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049718 100 mg
30050720 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049718 Leverantör Mikromol Leverantörsnummer MM0911.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[(1RS)-8-Ethyl-1-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (1-Propyl Etodolac)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[(1RS)-8-Ethyl-1-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (1-Propyl Etodolac)
CAS 57816-83-8
Analyt- eller komponentnamn 2-[(1RS)-8-Ethyl-1-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid (1-Propyl Etodolac)
Frakt skick Room Temperature
Molekylformel C18 H23 N O3
Certifikat och efterlevnad ISO 9001
Synonym 2-[(1RS)-8-Ethyl-1-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indol-1-yl]acetic Acid, Etodolac Imp. G (EP), 1-Propyl Etodolac, Etodolac Impurity G
InChI formel InChI=1S/C18H23NO3/c1-3-9-18(11-15(20)21)17-14(8-10-22-18)13-7-5-6-12(4-2)16(13)19-17/h5-7,19H,3-4,8-11H2,1-2H3,(H,20,21)
LEDER CCCC1(CC(=O)O)OCCc2c1[nH]c3c(CC)cccc23
IUPAC-namn 2-(8-ethyl-1-propyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetic acid
Molekylvikt (g/mol) 301.38
Kvantitet 100 mg
Formel vikt 301.1678 g/mol