Läs mer
2-[(1S,2S)-1-Ethyl-2-hydroxypropyl]-2,4-dihydro-4-[4-[4-(4-hydroxyphenyl)-1-piperazinyl]phenyl]-3H-1,2,4-triazol-3-one, TRC
Produktkod. 30376158
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376158 50 mg
30370861 10 mg
30372308 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30376158 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE68961050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 687132-01-0
Molekylformel C23H29N5O3
Molekylvikt (g/mol) 423.51
Synonym 1-((2S,3S)-2-Hydroxypentan-3-yl)-4-(4-(4-(4-hydroxyphenyl)piperazin-1-yl)phenyl)-1H-1,2,4-triazol-5(4H)-one
LEDER CC[C@@H]([C@H](C)O)N1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(O)cc4
Specifikationer
Kemiskt namn eller material 2-[(1S,2S)-1-Ethyl-2-hydroxypropyl]-2,4-dihydro-4-[4-[4-(4-hydroxyphenyl)-1-piperazinyl]phenyl]-3H-1,2,4-triazol-3-one
Kvantitet 50 mg
InChI formel InChI=1S/C23H29N5O3/c1-3-22(17(2)29)28-23(31)27(16-24-28)20-6-4-18(5-7-20)25-12-14-26(15-13-25)19-8-10-21(30)11-9-19/h4-11,16-17,22,29-30H,3,12-15H2,1-2H3/t17-,22-/m0/s1
Formel vikt 423.227