Läs mer
2-[2-[2-[2-[6-(Biotinylaminohexanoyl]aminoethoxy]ethoxy]ethoxy]-4-[3-(trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid N-Hydroxysuccinimide Ester, TRC
Produktkod. 30344081
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30344081 5 mg
1 options
Produktkod. 30344081 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3948665MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Benzoic acid, 2-[(21-[(3aR,4R,6aS)-hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-10,17-dioxo-3,6-dioxa-9,16-diazaheneicos-1-yl])oxy]-4-[3-(trifluoromethyl)-3H-diazirin-3-yl]-, 2,5-dioxo-1-pyrrolidinyl ester, rel-
CAS 1217722-17-2
Molekylformel C35H46F3N7O10S
Kvantitet 5 mg
Synonym 2-[2-[2-[2-[6-(Biotinylaminohexanoyl]aminoethoxy]ethoxy]ethoxy]-4-[3-(trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid N-Hydroxysuccinimide Ester
InChI formel InChI=1S/C35H46F3N7O10S/c36-35(37,38)34(43-44-34)22-9-10-23(32(50)55-45-29(48)11-12-30(45)49)25(20-22)54-19-18-53-17-16-52-15-14-40-28(47)7-2-1-5-13-39-27(46)8-4-3-6-26-31-24(21-56-26)41-33(51)42-31/h9-10,20,24,26,31H,1-8,11-19,21H2,(H,39,46)(H,40,47)(H2,41,42,51)/t24-,26-,31-/m0/s1
LEDER O=C1N[C@@H]2[C@@H]([C@H](CCCCC(NCCCCCC(NCCOCCOCCOC3=C(C(ON4C(CCC4=O)=O)=O)C=CC(C5(N=N5)C(F)(F)F)=C3)=O)=O)SC2)N1
IUPAC-namn (2,5-dioxopyrrolidin-1-yl) 2-[2-[2-[2-[6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]hexanoylamino]ethoxy]ethoxy]ethoxy]-4-[3-(trifluoromethyl)diazirin-3-yl]benzoate
Molekylvikt (g/mol) 813.84
Formel vikt 813.2979