Learn More
2-(2,2,2-Trifluoroethoxy)ethanol, TRC
Product Code. 30456852
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30456852 50 mg 50mg
1 options
Product Code. 30456852 Supplier Toronto Research Chemicals Supplier No. TRCT79558350MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(2,2,2-Trifluoroethoxy)ethanol
CAS 2358-54-5
Molecular Formula C4H7O2F3
Quantity 50 mg
InChI Formula InChI=1S/C4H7F3O2/c5-4(6,7)3-9-2-1-8/h8H,1-3H2
SMILES OCCOCC(F)(F)F
IUPAC Name 2-(2,2,2-trifluoroethoxy)ethanol
Molecular Weight (g/mol) 144.09
Formula Weight 144.0398