Learn More
2-[2-(2-Furyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, ≥97%, Thermo Scientific™
Product Code. 11310637
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11310637 250 mg Amber Glass Bottle 250mg
10081284 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11310637 Supplier Thermo Scientific Maybridge Supplier No. CC46239CB
 Restricted Item
Beräknad leverans: 05-01-2027
Add to basket
Chemical Identifiers
CAS 876316-28-8
Molecular Formula C16H19BO3
Molecular Weight (g/mol) 270.14
MDL Number MFCD08435879
InChI Key IKVHPKPYKOEGCK-UHFFFAOYSA-N
Synonym 2-2-fur-2-yl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-furan-2-yl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-2-furanyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-2-furyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 18525792
IUPAC Name 2-[2-(furan-2-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC=C1C1=CC=CO1
Specifications
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 270.13
Percent Purity 97%
Chemical Name or Material 2-[2-(2-Furyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane