Läs mer
2,2,2-Trifluoro-1-(3-hydroxypiperidin-1-yl)ethan-1-one (>90%), TRC
Produktkod. 30410617
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30410617 100 mg
30408983 500 mg
30424488 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30410617 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT206786100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73193-62-1
Molekylformel C7H10F3NO2
Molekylvikt (g/mol) 197.16
IUPAC-namn 2,2,2-trifluoro-1-(3-hydroxypiperidin-1-yl)ethanone
LEDER OC1CCCN(C1)C(=O)C(F)(F)F
Specifikationer
Kemiskt namn eller material 2,2,2-Trifluoro-1-(3-hydroxypiperidin-1-yl)ethan-1-one
Kvantitet 100 mg
InChI formel InChI=1S/C7H10F3NO2/c8-7(9,10)6(13)11-3-1-2-5(12)4-11/h5,12H,1-4H2
Formel vikt 197.0664