Läs mer
2,2,2-Trifluoro-N-phenylacetimidoyl Chloride, TRC
Produktkod. 30454175
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30454175 500 mg
1 options
Produktkod. 30454175 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT792605500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,2,2-Trifluoro-N-phenylacetimidoyl Chloride
CAS 61881-19-4
Molekylformel C8 H5 Cl F3 N
Kvantitet 500 mg
Synonym N-Phenyl-2,2,2-trifluoroacetimidoyl Chloride, N-Phenyltrifluoroacetimidoyl Chloride, 2,2,2-Trifluoro-N-phenylethanimidoyl Chloride
InChI formel InChI=1S/C8H5ClF3N/c9-7(8(10,11)12)13-6-4-2-1-3-5-6/h1-5H
LEDER FC(F)(F)C(=Nc1ccccc1)Cl
IUPAC-namn 2,2,2-trifluoro-N-phenylethanimidoyl chloride
Molekylvikt (g/mol) 207.58
Formel vikt 207.006