missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroacetamidine, 85%, Thermo Scientific™

Catalog Number 15214388
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
354-37-0
C2H3F3N2
112.06
MFCD00041545
NITMACBPVVUGOJ-UHFFFAOYSA-N
trifluoroacetamidine, trifluoroethanimidamide, 2,2,2-trifluoroacetamidine, ethanimidamide, 2,2,2-trifluoro, sntljqhjptadtaetp@, trifluoroacetoamidine, trifluoroacetamideine, trifluoro-acetamidine, acmc-20aje4, 2,2,2-trifluoroacetoamidine
2776882
2,2,2-trifluoroethanimidamide
NC(=N)C(F)(F)F
This item is not returnable. View return policy
354-37-0
C2H3F3N2
112.06
MFCD00041545
NITMACBPVVUGOJ-UHFFFAOYSA-N
trifluoroacetamidine, trifluoroethanimidamide, 2,2,2-trifluoroacetamidine, ethanimidamide, 2,2,2-trifluoro, sntljqhjptadtaetp@, trifluoroacetoamidine, trifluoroacetamideine, trifluoro-acetamidine, acmc-20aje4, 2,2,2-trifluoroacetoamidine
2776882
2,2,2-trifluoroethanimidamide
NC(=N)C(F)(F)F
Boiling Point 35.0°C to 37.0°C (11.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 84% min. (HPLC)
Quantity 5 g
Formula Weight 112.05
Percent Purity 85%
Chemical Name or Material 2,2,2-Trifluoroacetamidine
compliance-icons
Product Identifier
  • 2,2,2-Trifluoroacetamidine, 85%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'