missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl perfluorobutanesulphonate, TRC

Product Code. 30449285
Change view
Click to view available options
Kvantitet:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30449285 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30449285 Supplier Toronto Research Chemicals Supplier No. TRCT7905755G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2,2,2-Trifluoroethyl perfluorobutanesulphonate
CAS 79963-95-4
Molekylformel C6H2F12O3S
Kvantitet 5 g
Synonym 1,1,2,2,3,3,4,4,4-Nonafluorobutane-1-sulfonic Acid 2,2,2-trifluoroethyl Ester, 2,2,2-Trifluoroethyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonate, 2,2,2-Trifluoroethyl Nonafluorobutanesulfonate
InChI formel InChI=1S/C6H2F12O3S/c7-2(8,9)1-21-22(19,20)6(17,18)4(12,13)3(10,11)5(14,15)16/h1H2
LEDER FC(F)(F)COS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC-namn 2,2,2-trifluoroethyl 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonate
Molekylvikt (g/mol) 382.12
Formel vikt 381.9533
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.