Läs mer
2,2,3,3-Tetramethylsuccinimide, TRC
Produktkod. 30135269
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30135269 250 mg -
30008331 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135269 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT305330250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3566-61-8
Molekylformel C8 H13 N O2
Molekylvikt (g/mol) 155.19
Synonym 2,5-Pyrrolidinedione, 3,3,4,4-tetramethyl-, Succinimide, 2,2,3,3-tetramethyl- (6CI,7CI,8CI), 3,3,4,4-Tetramethyl-2,5-pyrrolidinedione, 2,2,3,3-Tetramethylsuccinimide, NSC 74524, PM 1090
IUPAC-namn 3,3,4,4-tetramethylpyrrolidine-2,5-dione
LEDER CC1(C)C(=O)NC(=O)C1(C)C
Specifikationer
Kemiskt namn eller material 2,2,3,3-Tetramethylsuccinimide
Anteckningar om renhetsgrad GC
Kvantitet 250 mg
InChI formel InChI=1S/C8H13NO2/c1-7(2)5(10)9-6(11)8(7,3)4/h1-4H3,(H,9,10,11)
Formel vikt 155.0946
Procent renhet >95%