Läs mer
2,2',3,4,4',5'-Hexabromodiphenyl Ether, TRC
Produktkod. 30406669
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30406669 50 mg
1 options
Produktkod. 30406669 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH29073550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material PBDE 138 (2,2',3,4,4',5'-Hexabromodiphenyl Ether)
CAS 182677-30-1
Molekylformel C12 H4 Br6 O
Kvantitet 50 mg
Synonym PBDE No. 138, Benzene, 1,2,3-tribromo-4-(2,4,5-tribromophenoxy)-, 1,2,3-Tribromo-4-(2,4,5-tribromophenoxy)benzene, 2,2',3,4,4',5'-Hexabromodiphenyl ether, BDE 138, PBDE 138, PBDE 138 (2,2',3,4,4',5'-Hexabromodiphenyl Ether), Benzene, 1,2,3-tribromo-4-(2,4,5-tribromophenoxy)-, 1,2,3-Tribromo-4-(2,4,5-tribromophenoxy)benzene, 2,2',3,4,4',5'-Hexabromodiphenyl ether, 2,2',3,4,4',5'-HexaBDE, BDE 138, PBDE 138
InChI formel InChI=1S/C12H4Br6O/c13-5-1-2-9(12(18)11(5)17)19-10-4-7(15)6(14)3-8(10)16/h1-4H
LEDER Brc1cc(Br)c(Oc2ccc(Br)c(Br)c2Br)cc1Br
IUPAC-namn 1,2,3-tribromo-4-(2,4,5-tribromophenoxy)benzene
Molekylvikt (g/mol) 643.58
Formel vikt 637.5362