missing translation for 'onlineSavingsMsg'
Läs mer

2-(2,3,4,6-Tetra-O-acetyl-Beta-D-galactopyranosyl)thiopseudourea Hydrobromide, TRC

Produktkod. 30424483
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30424483 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424483 Supplier Toronto Research Chemicals Supplier No. TRCT2796755MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 2-(2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranosyl)thiopseudourea Hydrobromide
CAS 51224-13-6
Molekylformel C15 H22 N2 O9 S . Br H
Kvantitet 5 mg
Synonym 2-(2,3,4,6-Tetra-O-Acetyl-B-D-Galactopyranosyl)Thiopseudourea Hydrobromide
InChI formel InChI=1S/C15H22N2O9S.BrH/c1-6(18)22-5-10-11(23-7(2)19)12(24-8(3)20)13(25-9(4)21)14(26-10)27-15(16)17;/h10-14H,5H2,1-4H3,(H3,16,17);1H/t10-,11+,12+,13-,14+;/m1./s1
LEDER Br.CC(=O)OC[C@H]1O[C@@H](SC(=N)N)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC-namn [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-carbamimidoylsulfanyloxan-2-yl]methyl acetate;hydrobromide
Molekylvikt (g/mol) 487.32
Formel vikt 486.0308
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.