Läs mer
2,2'-[[[4-(3-Oxo-4-morpholinyl)phenyl]imino]bis[(2R)-2-hydroxy-3,1-propanediyl]]bis[1H-isoindole-1,3(2H)-dione], TRC
Produktkod. 30427948
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427948 250 mg
30424860 50 mg
30420211 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427948 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO858058250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1643354-27-1
Molekylformel C32H30N4O8
Molekylvikt (g/mol) 598.6
Synonym 1H-Isoindole-1,3(2H)-dione, 2,2'-[[[4-(3-oxo-4-morpholinyl)phenyl]imino]bis[(2R)-2-hydroxy-3,1-propanediyl]]bis- (ACI), 2,2'-[[[4-(3-Oxo-4-morpholinyl)phenyl]imino]bis[(2R)-2-hydroxy-3,1-propanediyl]]bis[1H-isoindole-1,3(2H)-dione] (ACI), 2,2'-[(2R,2'R)-[[4-(3-oxomorpholino)phenyl]azanediyl]bis(2-hydroxypropane-3,1-diyl)]bis(isoindoline-1,3-dione)
IUPAC-namn 2-[(2R)-3-[N-[(2R)-3-(1,3-dioxoisoindol-2-yl)-2-hydroxypropyl]-4-(3-oxomorpholin-4-yl)anilino]-2-hydroxypropyl]isoindole-1,3-dione
LEDER O=C1N(C2=CC=C(N(C[C@@H](O)CN(C(C3=C4C=CC=C3)=O)C4=O)C[C@@H](O)CN(C(C5=C6C=CC=C5)=O)C6=O)C=C2)CCOC1
Specifikationer
Kemiskt namn eller material 2,2'-[((4-(3-Oxo-4-morpholinyl)phenyl]imino])bis[(2R)-2-hydroxy-3,1-propanediyl)]bis[1H-isoindole-1,3(2H)-dione]
Kvantitet 250 mg
InChI formel InChI=1S/C32H30N4O8/c37-22(17-35-29(40)24-5-1-2-6-25(24)30(35)41)15-33(20-9-11-21(12-10-20)34-13-14-44-19-28(34)39)16-23(38)18-36-31(42)26-7-3-4-8-27(26)32(36)43/h1-12,22-23,37-38H,13-19H2/t22-,23-/m1/s1
Formel vikt 598.2064