Läs mer
2,2',4,4',5,5'-Hexabromodiphenyl Ether, TRC
Produktkod. 30133040
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133040 100 mg
17934695 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133040 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH290730100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 68631-49-2
Molekylformel C12 H4 Br6 O
Molekylvikt (g/mol) 643.58
Synonym Benzene, 1,1'-oxybis[2,4,5-tribromo-, 1,1'-Oxybis[2,4,5-tribromobenzene], 2,2',4,4',5,5'-Hexabromodiphenyl ether, BDE 153, PBDE 153, PBDE No. 153
IUPAC-namn 1,2,4-tribromo-5-(2,4,5-tribromophenoxy)benzene
LEDER Brc1cc(Br)c(Oc2cc(Br)c(Br)cc2Br)cc1Br
Specifikationer
Kemiskt namn eller material PBDE 153 (2,2',4,4',5,5'-Hexabromodiphenyl Ether)
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H4Br6O/c13-5-1-9(17)11(3-7(5)15)19-12-4-8(16)6(14)2-10(12)18/h1-4H
Formel vikt 637.5362
Procent renhet >95%