Läs mer
2,2,4,4,6,6-Hexakis-phenyl-1,3,5,2,4,6-trioxatrisilinane, TRC
Produktkod. 30179904
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179904 100 mg
30180410 250 mg
30180174 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179904 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH291018100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 512-63-0
Molekylformel C36 H30 O3 Si3
Molekylvikt (g/mol) 594.878
Synonym hexaphenyl-Cyclotrisiloxane, 2,2,4,4,6,6-Hexaphenylcyclotrisiloxane, Diphenylsiloxane cyclic trimer, Hexaphenylcyclotrisiloxane, NSC 127080
IUPAC-namn 2,2,4,4,6,6-hexakis-phenyl-1,3,5,2,4,6-trioxatrisilinane
LEDER O1[Si](O[Si](O[Si]1(c2ccccc2)c3ccccc3)(c4ccccc4)c5ccccc5)(c6ccccc6)c7ccccc7
Specifikationer
Kemiskt namn eller material 2,2,4,4,6,6-Hexakis-phenyl-1,3,5,2,4,6-trioxatrisilinane
Kvantitet 100 mg
InChI formel InChI=1S/C36H30O3Si3/c1-7-19-31(20-8-1)40(32-21-9-2-10-22-32)37-41(33-23-11-3-12-24-33,34-25-13-4-14-26-34)39-42(38-40,35-27-15-5-16-28-35)36-29-17-6-18-30-36/h1-30H
Formel vikt 594.15