Läs mer
2,2',4,4',6-Pentabromodiphenyl Ether, TRC
Produktkod. 30133591
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133591 100 mg
17981585 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133591 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP237820100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 189084-64-8
Molekylformel C12 H5 Br5 O
Molekylvikt (g/mol) 564.69
Synonym Benzene, 1,3,5-tribromo-2-(2,4-dibromophenoxy)-, 1,3,5-Tribromo-2-(2,4-dibromophenoxy)benzene, 2,2',4,4',6-Pentabromodiphenyl ether, BDE 100, PBDE 100
IUPAC-namn 1,3,5-tribromo-2-(2,4-dibromophenoxy)benzene
LEDER Brc1ccc(Oc2c(Br)cc(Br)cc2Br)c(Br)c1
Specifikationer
Kemiskt namn eller material PBDE 100 (2,2',4,4',6-Pentabromodiphenyl Ether)
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H5Br5O/c13-6-1-2-11(8(15)3-6)18-12-9(16)4-7(14)5-10(12)17/h1-5H
Formel vikt 559.6257
Procent renhet >95%