Läs mer
2,2',4,4'-Tetranitro-1,1'-biphenyl, TRC
Produktkod. 30453904
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30453904 500 mg
30449775 100 mg
30449579 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30453904 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT305780500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1820-59-3
Molekylformel C12H6N4O8
Molekylvikt (g/mol) 334.2
Synonym 2,2',4,4'-Tetranitrobiphenyl, 2,4,2',4'-Tetranitrobiphenyl, NSC 97248
LEDER NC1Cc2c(C1)cc3c(OCO3)c2
Specifikationer
Kemiskt namn eller material 2,2',4,4'-Tetranitro-1,1'-biphenyl
Kvantitet 500 mg
InChI formel InChI=1S/C12H6N4O8/c17-13(18)7-1-3-9(11(5-7)15(21)22)10-4-2-8(14(19)20)6-12(10)16(23)24/h1-6H