Läs mer
2-[2-[[4-[(5S)-5-[[[(5-Chloro-2-thienyl)carbonyl]amino]methyl]-2-oxo-3-oxazolidinyl]phenyl]amino]ethoxy]-acetic Acid Hydrochloride, TRC
Produktkod. 30351246
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30351246 25 mg
1 options
Produktkod. 30351246 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC42184025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[2-[(4-[(5S)-5-[([(5-Chloro-2-thienyl)carbonyl]amino)methyl]-2-oxo-3-oxazolidinyl)phenyl]amino]ethoxy]-acetic Acid Hydrochloride
CAS 931117-61-2
Molekylformel C19H20ClN3O6S.HCl
Kvantitet 25 mg
Synonym Acetic acid, 2-[2-[[4-[(5S)-5-[[[(5-chloro-2-thienyl)carbonyl]amino]methyl]-2-oxo-3-oxazolidinyl]phenyl]amino]ethoxy]-, hydrochloride (1:1) (ACI), 2-[2-[[4-[(5S)-5-[[[(5-Chloro-2-thienyl)carbonyl]amino]methyl]-2-oxo-3-oxazolidinyl]phenyl]amino]ethoxy]acetic Acid Hydrochloride
InChI formel InChI=1S/C19H20ClN3O6S.ClH/c20-16-6-5-15(30-16)18(26)22-9-14-10-23(19(27)29-14)13-3-1-12(2-4-13)21-7-8-28-11-17(24)25;/h1-6,14,21H,7-11H2,(H,22,26)(H,24,25);1H/t14-;/m0./s1
LEDER Cl.OC(=O)COCCNc1ccc(cc1)N2C[C@H](CNC(=O)c3ccc(Cl)s3)OC2=O
IUPAC-namn 2-[2-[4-[(5S)-5-[[(5-chlorothiophene-2-carbonyl)amino]methyl]-2-oxo-1,3-oxazolidin-3-yl]anilino]ethoxy]acetic acid;hydrochloride
Molekylvikt (g/mol) 490.3575
Formel vikt 489.0528