missing translation for 'onlineSavingsMsg'
Läs mer

2,2'-((4-Aminophenyl)azanediyl)diethanol Sulfate, TRC

Produktkod. 30330347
Change view
Click to view available options
Kvantitet:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30330347 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330347 Supplier Toronto Research Chemicals Supplier No. TRCA6381431G

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N,N-Bis(2-hydroxyethyl)-1,4-phenylenediamine Sulfate
CAS 54381-16-7
Molekylformel C10 H16 N2 O2 . H2 O4 S
Kvantitet 1 g
Synonym 2,2'-[(4-Aminophenyl)imino]bisethanol sulfate, 4-Amino-N,N-di(β-hydroxyethyl)aniline sulfate, N,N-Bis(2-hydroxy)-p-phenylenediamine sulfate, N,N-Bis(2-hydroxyethyl)-p-phenylenediamine sulfate, N,N-Bis(hydroxyethyl)-p-phenylenediamine sulfate, N,N-Bis(β-hydroxyethyl)-p-phenylenediamine monosulfate, Ro 371, N,N-Bis(2-hydroxyethyl)-1,4-phenylenediamine sulfate, N,N-Bis(2-hydroxyethyl)-1,4-phenylenediamine Sulfate
InChI formel InChI=1S/C10H16N2O2.H2O4S/c11-9-1-3-10(4-2-9)12(5-7-13)6-8-14;1-5(2,3)4/h1-4,13-14H,5-8,11H2;(H2,1,2,3,4)
LEDER Nc1ccc(cc1)N(CCO)CCO.OS(=O)(=O)O
IUPAC-namn 2-[4-amino-N-(2-hydroxyethyl)anilino]ethanol;sulfuric acid
Molekylvikt (g/mol) 294.3247
Formel vikt 294.0886
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.