Läs mer
2-(2-(4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl)ethoxy)ethyl AcetateQuetiapine Acetate, TRC
Produktkod. 30351067
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30351067 250 mg
1 options
Produktkod. 30351067 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD417095250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Quetiapine Impurity B
CAS 844639-07-2
Molekylformel C23 H27 N3 O3 S
Kvantitet 250 mg
Synonym 2-(2-(4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl)ethoxy)ethyl Acetate Quetiapine Acetate, 11-[4-[2-(2-Acetoxyethoxy)ethyl]piperazin-1-yl]dibenzo[b,f][a,4]thiazepine, 2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]-ethanol Acetate, Quetiapine USP-B
InChI formel InChI=1S/C23H27N3O3S/c1-18(27)29-17-16-28-15-14-25-10-12-26(13-11-25)23-19-6-2-4-8-21(19)30-22-9-5-3-7-20(22)24-23/h2-9H,10-17H2,1H3
LEDER CC(=O)OCCOCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24
IUPAC-namn 2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethyl acetate
Molekylvikt (g/mol) 425.5438
Formel vikt 425.1773