Läs mer
2-(2,4-Dichlorophenyl)-oxirane, TRC
Produktkod. 30383918
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30383918 50 mg
30372777 10 mg
30381576 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30383918 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47568350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13692-15-4
Molekylformel C8H6OCl2
Molekylvikt (g/mol) 189.03
Synonym Oxirane, 2-(2,4-dichlorophenyl)- (ACI), 2-(2,4-Dichlorophenyl)oxirane (ACI), Benzene, 2,4-dichloro-1-(epoxyethyl)- (8CI), Oxirane, (2,4-dichlorophenyl)- (9CI), (2,4-Dichlorophenyl)oxirane
IUPAC-namn 2-(2,4-dichlorophenyl)oxirane
LEDER Clc1ccc(C2CO2)c(Cl)c1
Specifikationer
Kemiskt namn eller material 2-(2,4-Dichlorophenyl)-oxirane
Kvantitet 50 mg
InChI formel InChI=1S/C8H6Cl2O/c9-5-1-2-6(7(10)3-5)8-4-11-8/h1-3,8H,4H2
Formel vikt 187.9796