missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(2,5-Difluorophenyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64215.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15451319

  • 10248.91 SEK / 5g


Estimated Shipment date
from Supplier 14-11-2018
Add to basket

Specifications

Chemical Identifiers

MFCD11645340

Specifications

2-(2,5-Difluorophenyl)benzimidazole
MFCD11645340
Solid
5g
230.22
95%
Videos
SDS