Learn More
2-[(2,6-Dichlorophenyl)amino]benzaldehyde (Diclofenac impurity), TRC
Product Code. 30136726
Change view
Click to view available options
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30136726 25 mg
30009489 5 mg
30012780 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136726 Supplier Toronto Research Chemicals Supplier No. TRCD43566025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22121-58-0
Molekylformel C13 H9 Cl2 N O
Molekylvikt (g/mol) 266.12
Synonym Anthranilaldehyde, N-(2,6-dichlorophenyl)- (8CI), 2-[(2,6-Dichlorophenyl)amino]benzaldehyde, 2-(2,6-Dichloroanilino)benzaldehyde, o-(2,6-Dichloroanilino)benzaldehyde
IUPAC-namn 2-(2,6-dichloroanilino)benzaldehyde
LEDER Clc1cccc(Cl)c1Nc2ccccc2C=O
Specifications
Kemiskt namn eller material 2-[(2,6-Dichlorophenyl)amino]benzaldehyde
Kvantitet 25 mg
InChI formel InChI=1S/C13H9Cl2NO/c14-10-5-3-6-11(15)13(10)16-12-7-2-1-4-9(12)8-17/h1-8,16H
Formel vikt 265.0061