Läs mer
2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Benzoate, Mikromol
Produktkod. 30461263
Change view
Kvantitet:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30461263 4 x 25 mg
30050165 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30461263 Leverantör Mikromol Leverantörsnummer MM0061.0400254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Benzoate, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Benzoate
CAS 59277-91-7
Analyt- eller komponentnamn 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Benzoate
Frakt skick Room Temperature
Molekylformel C15 H15 N5 O4
Certifikat och efterlevnad ISO 17025
Synonym 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl Benzoate
InChI formel InChI=1S/C15H15N5O4/c16-15-18-12-11(13(21)19-15)17-8-20(12)9-23-6-7-24-14(22)10-4-2-1-3-5-10/h1-5,8H,6-7,9H2,(H3,16,18,19,21)
Lösningstyp Neat
LEDER NC1=Nc2c(ncn2COCCOC(=O)c3ccccc3)C(=O)N1
IUPAC-namn 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethyl benzoate
Molekylvikt (g/mol) 329.31
Kvantitet 4 x 25 mg
Formel vikt 329.1124 g/mol