Learn More
2-(2-Aminobenzoyl)pyridine, TRC
Product Code. 30320411
Change view
Click to view available options
Kvantitet:
1 g
Product Code. Kvantitet
30320411 1 g
1 options
Product Code. 30320411 Supplier Toronto Research Chemicals Supplier No. TRCA5938051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2-(2-Aminobenzoyl)pyridine
CAS 42471-56-7
Molekylformel C12 H10 N2 O
Kvantitet 1 g
Synonym (2-Aminophenyl)-2-pyridinylmethanone, (2-Aminophenyl)pyridin-2-ylmethanone, 2-Aminophenyl 2-pyridyl Ketone
InChI formel InChI=1S/C12H10N2O/c13-10-6-2-1-5-9(10)12(15)11-7-3-4-8-14-11/h1-8H,13H2
LEDER Nc1ccccc1C(=O)c2ccccn2
IUPAC-namn (2-aminophenyl)-pyridin-2-ylmethanone
Molekylvikt (g/mol) 198.22
Formel vikt 198.0793